Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch4 Hamster anti-Mouse, Biotin, Clone: HMN4-14, BD Phrmingen™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 569021
Description
Notch4 Monoclonal antibody specifically detects Notch4 in Mouse samples. It is validated for Flow cytometry
Specifications
Notch4 | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
18132 | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with biotin under optimum conditions, and unreacted biotin was removed. | |
IgG κ |
Flow Cytometry | |
HMN4-14 | |
Biotin | |
Notch 4; Notch gene homolog 4; Notch4 | |
Mouse Extracellular Notch4 Domain Recombinant Protein | |
0.1 mg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction