Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOTCH4 Monoclonal Antibody (HMN4-14), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010852
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-108-52 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-108-52 Supplier Invitrogen™ Supplier No. 12576480
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: This HMN4-14 monoclonal antibody reacts with mouse Notch4, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, forms a ternary complex with the DNA-binding transcription factor CSL (CBF-1, Su(H), Lag-1) and the coactivator Mastermind, and ultimately activates transcription. Notch 4 has been shown to be expressed in CD8+ splenic dendritic cells, endothelial cells (e.g. vascular smooth muscle cells), and macrophages. A truncated, constitutively active form of Notch 4 designated Int3 also exists in mice as a result of the mouse mammary tumor virus (MMTV) insertion into the Notch 4 gene. Composed of the transmembrane and intracellular domains of Notch 4, Int3 has been reported to be involved in mammary gland development and tumorigenesis in the mouse. Finally, studies have shown Notch 4 involvement in myeloid proliferation, hematopoiesis, and embryonic endothelial development. The HMN4-14 antibody is reported to also crossreact to rat and...

This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases (entrez gene).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOTCH4
Applications Flow Cytometry
Classification Monoclonal
Clone HMN4-14
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene NOTCH4
Gene Accession No. P31695
Gene Alias DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3
Gene Symbols NOTCH4
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18132, 406162
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.