Learn More
Invitrogen™ NOTCH4 Polyclonal Antibody

Description
Peptide sequence: KALKPKAEVD EDGVVMCSGP EEGEEVGQAE ETGPPSTCQL WSLSGGCGAL Sequence homology: Cow: 92%; Dog: 77%; Guinea Pig: 79%; Horse: 92%; Human: 100%; Pig: 100%; Rabbit: 79%.
Specifications
Specifications
| Antigen | NOTCH4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | NOTCH4 |
| Gene Accession No. | Q99466 |
| Gene Alias | DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3 |
| Gene Symbols | NOTCH4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.