Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOTCH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115664
Description
Antibody detects endogenous levels of total NOTCH4.
This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases (entrez gene).
Specifications
| NOTCH4 | |
| Polyclonal | |
| Unconjugated | |
| NOTCH4 | |
| DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18132, 406162, 4855 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P31695, Q99466 | |
| NOTCH4 | |
| A synthesized peptide derived from human NOTCH4(Accession Q99466), corresponding to amino acid residues R1474-G1524. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction