Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ notchl Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117530
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117530 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117530 Supplier Invitrogen™ Supplier No. PA5117530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

notchl is predicted to be involved in the Notch signaling pathway, cell differentiation, and multicellular organism development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen notchl
Applications Western Blot
Classification Polyclonal
Concentration 2.509 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene notchl
Gene Accession No. 0
Gene Alias neurogenic locus notch homolog protein 1; Notch homolog; notch homolog, like; notch receptor, like; notchl
Gene Symbols notchl
Host Species Rabbit
Immunogen Recombinant Zebrafish Notch homolog,-like protein (683-784aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 569522
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.