Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Noto Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549419
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549419 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549419 Supplier Invitrogen™ Supplier No. PA549419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Noto Polyclonal Antibody for Western Blot

Transcription regulator acting downstream of both FOXA2 and T during notochord development. Required for node morphogenesis. Is essential for cilia formation in the posterior notochord (PNC) and for left-right patterning; acts upstream of FOXJ1 and RFX3 in this process and is required for the expression of various components important for axonemal assembly and function. Plays a role in regulating axial versus paraxial cell fate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Noto
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Noto
Gene Accession No. Q5TIS6
Gene Alias Flh; floating head homolog; homeobox protein notochord; MmNot; Not; Not homeodomain protein; NOTO; notochord homeobox; notochord homolog; notochord homolog (Xenopus laevis); tc; truncate
Gene Symbols Noto
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 200-234 amino acids from the C-terminal region of Mouse Noto.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 384452
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.