Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Noto Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143403
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Transcription regulator acting downstream of both FOXA2 and T during notochord development. Required for node morphogenesis. Is essential for cilia formation in the posterior notochord (PNC) and for left-right patterning; acts upstream of FOXJ1 and RFX3 in this process and is required for the expression of various components important for axonemal assembly and function. Plays a role in regulating axial versus paraxial cell fate.
Specifications
| Noto | |
| Polyclonal | |
| Unconjugated | |
| Noto | |
| Flh; floating head homolog; homeobox protein notochord; MmNot; Not; Not homeodomain protein; NOTO; notochord homeobox; notochord homolog; notochord homolog (Xenopus laevis); tc; truncate | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30260 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| 0 | |
| Noto | |
| Recombinant protein encompassing a sequence within the center region of zebrafish Flh. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction