Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTUM Antibody, Novus Biologicals™
SDP

Catalog No. NBP266631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266631 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266631 Supplier Novus Biologicals Supplier No. NBP266631
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOTUM Polyclonal specifically detects NOTUM in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NOTUM
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:10 - 1:500
Formulation PBS with 0.08% Sodium Azide
Gene Alias EC 3.-, notum pectinacetylesterase homolog (Drosophila), protein notum homolog
Gene Symbols NOTUM
Host Species Rabbit
Immunogen Rabbits were immunized with recombinant full-length notum protein from mouse.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 147111
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.