Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOTUM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113025
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113025 Supplier Invitrogen™ Supplier No. PA5113025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOTUM Polyclonal Antibody for Western Blot, ELISA

Genetic studies in Drosophila have revealed a novel secreted antagonist for Wingless (Wg) signaling, referred to as Notum/Wingful (Wf). Wf/Notum encodes a protein of 430 amino acid residues, which is related to plant pectin acetylesterases of the a/beta-hydrolase superfamily. The main target of Notum action appears to be cell surface proteoglycans Dally and/or Dally-like (Dlp). Notum governs the extracellular Wg morphogen distribution via Dally and Dlp. Also, like some vertebrate Wnt antagonists, notum expression is activated by Wg signaling, revealing the existence of a feedback control mechanism in which Wg modifies its own gradient distribution. In vertebrate, proteoglycans related to Dally and Dlp likely play important roles in Wnt signaling which encodes a glypican that is required for Wnt-11 signaling. A single ortholog of Notum exists in the mouse and the human genome, implying that a similar regulatory mechanism may exist in vertebrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOTUM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4
Gene NOTUM
Gene Accession No. Q6P988, Q8R116
Gene Alias [Wnt protein] O-palmitoleoyl-L-serine hydrolase; 5730593N15Rik; hNOTUM; NOTUM; notum pectinacetylesterase homolog; notum pectinacetylesterase homolog (Drosophila); NOTUM, palmitoleoyl-protein carboxylesterase; OK/SW-CL.30; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog; RGD1307119
Gene Symbols NOTUM
Host Species Rabbit
Immunogen Recombinant Human Palmitoleoyl-protein carboxylesterase NOTUM protein (100-235AA).
Purification Method Antigen Affinity Chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147111, 77583
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.