Learn More
Invitrogen™ NOTUM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595811
Description
Immunogen sequence: SEGRKTWRRR GQQPPPPPRT EAAPAAGQPV ESFPLDFTAV EGNMDSFMAQ VKSLAQSLYP CSAQQLNEDL RLHLLLNTSV TCNDGSPAGY YLKESRGSRR WLLFLEGGWY CFNRENCDSR YDTMRRLMSS RDWPRTRTGT GILSSQPEEN PYWWNANMVF IPYCSSDVWS GASSKSEKNE YAFMGALIIQ EVVRELLGRG LSGAKVLLLA GSSAGGTGVL LNVDRVAEQL EKLGYPAIQV RGLADSGWFL DNKQYRHTDC VDTITCAPTE AIRRGIRYWN GVVPERCRRQ FQEGEEWNCF F; Positive Samples: BxPC-3, HepG2.
Genetic studies in Drosophila have revealed a novel secreted antagonist for Wingless (Wg) signaling, referred to as Notum/Wingful (Wf). Wf/Notum encodes a protein of 430 amino acid residues, which is related to plant pectin acetylesterases of the a/beta-hydrolase superfamily. The main target of Notum action appears to be cell surface proteoglycans Dally and/or Dally-like (Dlp). Notum governs the extracellular Wg morphogen distribution via Dally and Dlp. Also, like some vertebrate Wnt antagonists, notum expression is activated by Wg signaling, revealing the existence of a feedback control mechanism in which Wg modifies its own gradient distribution. In vertebrate, proteoglycans related to Dally and Dlp likely play important roles in Wnt signaling which encodes a glypican that is required for Wnt-11 signaling. A single ortholog of Notum exists in the mouse and the human genome, implying that a similar regulatory mechanism may exist in vertebrates.
Specifications
NOTUM | |
Polyclonal | |
Unconjugated | |
NOTUM | |
[Wnt protein] O-palmitoleoyl-L-serine hydrolase; 5730593N15Rik; hNOTUM; NOTUM; notum pectinacetylesterase homolog; notum pectinacetylesterase homolog (Drosophila); NOTUM, palmitoleoyl-protein carboxylesterase; OK/SW-CL.30; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog; RGD1307119 | |
Rabbit | |
Affinity chromatography | |
RUO | |
147111 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.23 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6P988 | |
NOTUM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-320 of human NOTUM (NP_8485883). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.