Learn More
NOTUM Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NOTUM |
---|---|
Concentration | 333 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8334
![]() ![]() |
Proteintech Group Inc
146631AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The NOTUM antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NOTUM. This antibody recognizes human, mouse antigen. The NOTUM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NOTUM | |
Western Blot | |
Unconjugated | |
Human, Mouse | |
IgG |
333 μg/mL | |
Polyclonal | |
Rabbit | |
147111 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.