Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOV/CCN3 Antibody (130), Alexa Fluor™ 647 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289369AF647
Description
NOV/CCN3 Monoclonal antibody specifically detects NOV/CCN3 in Human samples. It is validated for ELISASpecifications
NOV/CCN3 | |
Monoclonal | |
Alexa Fluor 647 | |
CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human NOV/CCN3 (NP_002505.1; Met 1-Met 357). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
130 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
4856 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction