Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nova-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571356
Description
Nova-2 Polyclonal Antibody for Western Blot
Paraneoplastic opsoclonus myoclonus ataxia (POMA) is a neurologic disorder thought to be mediated by an autoimmune attack against onconeural disease antigens that are expressed by gynecologic or lung tumors and by neurons. Nova-1 and -2 are both POMA disease antigens. The autoimmune attack against Nova-2 in POMA patients results in latent cancer, reduced inhibitory control of movements, and dementia. Nova-2 is closely related to Nova-1, and is expressed at high levels in neurons during development and at lower levels in the adult lung. In the postnatal mouse brain, Nova-2 is expressed in a pattern that is largely reciprocal with Nova-1, including high levels of Nova-2 expression in the neocortex and hippocampus. Both Novas are RNA binding proteins, and Nova-2 binds RNA with high affinity and with sequence specificity that differs from Nova-1. They regulate neuronal pre-messenger RNA splicing by directly binding to RNA. Nova-2 target genes function either in the synapse or in axon guidance indicating that Nova-2 is a major regulator of synaptic function.
Specifications
Nova-2 | |
Polyclonal | |
Unconjugated | |
NOVA2 | |
ANOVA; astrocytic NOVA1-like RNA-binding protein; Gm1424; neuro-oncological ventral antigen 2; neuro-oncological ventral antigen 3; NOVA alternative splicing regulator 2; NOVA2; NOVA3; RNA-binding protein Nova-2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4858 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UNW9 | |
NOVA2 | |
synthetic peptide directed towards the middle region of human Nova-2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction