Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOVA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521459
Description
Recommended positive controls: PC-3. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Chicken (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described.
Specifications
| NOVA1 | |
| Polyclonal | |
| Unconjugated | |
| NOVA1 | |
| 9430099M15Rik; G630039L02; neuro-oncological ventral antigen 1; NOVA alternative splicing regulator 1; Nova1; Nova-1; Onconeural ventral antigen 1; paraneoplastic Ri antigen; RNA-binding protein Nova-1; ventral neuron-specific protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.92 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P51513 | |
| NOVA1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human NOVA1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction