Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOVA1 Polyclonal Antibody

Catalog No. PIPA5121737
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121737 20 μL
PIPA5119305 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121737 Supplier Invitrogen™ Supplier No. PA5121737
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOVA1 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOVA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NOVA1
Gene Accession No. P51513
Gene Alias 9430099M15Rik; G630039L02; neuro-oncological ventral antigen 1; NOVA alternative splicing regulator 1; Nova1; Nova-1; Onconeural ventral antigen 1; paraneoplastic Ri antigen; RNA-binding protein Nova-1; ventral neuron-specific protein 1
Gene Symbols NOVA1
Host Species Rabbit
Immunogen Recombinant protein Neuro Oncological Ventral Antigen 1. The antigen corresponds to amino acid range 292-507 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.