Learn More
Cytiva NovaBlot Kit
Catalog No. 45002382 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
Electrophoretic Transfer
Catalog No. For Use With (Application)
45-002-382 Electrophoretic Transfer
1 options
Catalog No. 45-002-382 Supplier Cytiva Supplier No. 18101686
Includes
1 x Electrode cathode, 1 x Electrode anode, 500 x Electrode paper (200 x 250mm), 50 x Cellophane sheet (210 x 320mm)
For electrophoretic transfer applications.
  • For use with Multiphor II system
  • Kit includes electrode cathode, anode, electrode paper, cellophane sheet
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Electrophoretic Transfer
Includes 1 x Electrode cathode, 1 x Electrode anode, 500 x Electrode paper (200 x 250mm), 50 x Cellophane sheet (210 x 320mm)
Quantity 1 Kit