Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ B834pLysS Competent Cells

Catalog No. 690423 Shop All MilliporeSigma Products
Click to view available options
Quantity:
0.4mL
1mL

For production of protein from target genes cloned in pET vectors by induction with IPTG

  • B834 is parental strain for BL21 (1)
  • Protease-deficient hosts are methionine auxotrophs
  • Allows high specific-activity labeling of target proteins with methionine and selenomethionine for crystallography
  • pLysS strains express T7 lysozyme
  • Suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤ -70°C
Product Type Competent Cells
Quantity 0.4mL

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.