Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ His Bind™ Resin
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mL
50 mL
Description
EMD Millipore Novagen™ His Bind™ Resin is an uncharged IDA agarose.
- Binding capacity is approximately 8mg/mL bed volume
- Uses either gravity flow columns or batch methods

Specifications
Specifications
Content And Storage | 2°C to 8°C |
Quantity | 10 mL |
Type | Chromatography Media |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction