Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ Insect GeneJuice™ Transfection Reagent
Click to view available options
Quantity:
10 mL
Description
EMD Millipore Novagen™ Insect GeneJuice™ Transfection Reagent used for both transient and stable transfections.
- Low toxicity to cells
- Used for transfection of transfer plasmids with linearized virus DNA
- For reproduction of recombinant baculovirus
- For large-scale protein expression in suspension-culture transfections of Sf9 insect cells
- Packaging: Glass Bottle
Applications:

Specifications
Specifications
Content And Storage | 2°C to -8°C |
Quantity | 10 mL |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction