Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ LB Broth, Miller

Catalog No. 717535 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Easy
5 kg
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-753-5 500 g
71-753-4 25 Easy
71-753-6 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-753-5 Supplier MilliporeSigma™ Supplier No. 717535
Add to Cart
Add to Cart

Used as granulated medium for cultivation of E. coli on scales ranging from small cultures to fermentation. MilliporeSigma™; Novagen™ LB Broth, Miller granules ensure rapid and uniform dissolution in water.

  • Prevents clumping of medium and inhalation of airborne powder
  • Supplied in several formats
  • EasyPak is aluminum foil pouch containing 25g granulated medium sufficient for 1L culture
  • Add EasyPak contents to 1L water, microwave for 2 minutes to dissolve agar and sterilize
  • Supplied in 500g and 5kg bottles
  • Composition of LB Agar MILLER per liter is 5g yeast extract, 10g peptone from casein, 10g sodium chloride
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Product Type Microbiology Media
Quantity 500 g

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.