Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Origami™ 2 (DE3) pLysS Competent Cells

Catalog No. 713463 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-346-3 0.4 mL
71-346-4 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-346-3 Supplier MilliporeSigma™ Supplier No. 713463
Add to Cart
Add to Cart

Origami 2 strains have mutations in glutathione reducatase(gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation. T7 lysozyme expression suppresses basal T7 expression.

Origami™ 2 host strains are K-12 derivatives that have mutations in both the thioredoxin reductase (trxB) and glutathione reductase (gor) genes, which greatly enhance disulfide bond formation in the E. coli cytoplasm. The Origami 2 strains are kanamycin sensitive; like the original Origami strains, the gor mutation is still selected for by tetracycline. To reduce the possibility of disulfide bond formation between molecules, strains containing mutations in trxB and gor are recommended only for the expression of proteins that require disulfide bond formation for proper folding. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.