Learn More
Description
- Instant TB Medium is combination of Overnight Express Autoinduction System 1 components (Cat. No. 71300) with rich Terrific Broth (TB) medium and is supplied in two formats
- EasyPak is aluminum foil pouch containing 60g granulated medium, sufficient for 1L culture
- Add the EasyPak contents to 1L sterile water, supplement with 10mL glycerol, and microwave for 2 min to dissolve
- Available in 1kg bottles
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Product Type | Microbiology Media |
| For Use With (Application) | Protein production |
| Packaging Type | Glass Bottle |
| Quantity | 5 Easy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.