Learn More
MilliporeSigma™ Novagen™ pSTBlue-1 AccepTor™ Vector Kit
Catalog No. p-6921050
Click to view available options
Quantity:
20 Reactions
40 Reactions
Designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases, which leave single 3'-dA overhangs on the reaction products. MilliporeSigma™; Novagen™ pSTBlue-1 AccepTor™ Vector Kit are supplied in a ready-to-ligate format.
  • No restriction enzymes or special primers
  • Direct ligation of PCR product with vector
  • Compatible with polymerases that leave single 3´-dA overhangs
  • Blue/white screening with pSTBlue-1 or pETBlue™-1 vectors
  • Simple protocol takes as little as 40 minutes from PCR product to plating transformants
TRUSTED_SUSTAINABILITY
Specifications
Product Type pSTBlue-1 Vector Kit
Content And Storage Store at ≤70°C.
Quantity 20 Reactions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.