Learn More
Description
- Contains highly purified and stabilized recombinant lysozyme
- Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
- Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
- Optimally active at physiological pH
Specifications
Specifications
| Concentration | 27 to 33K U/μL |
| Components | Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100 |
| pH | 7.5 |
| Content And Storage | -20°C |
| Quantity | 300 kU |
| Product Type | Stabilized recombinant lysozyme |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.