Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NovaLume Atto Chemiluminescent Substrate (AP)
SDP

Catalog No. NBP261914 Shop All Novus Products
Change view
Click to view available options
Packaging Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
NBP2-61914 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61914 Supplier R&D Systems™ Supplier No. NBP261914

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. NovaLume Atto Chemiluminescent Substrate (AP) is a ready-to-use stabilized 1, 2-dioxetane substrate.

  • Extreme sensitivity, from low nano to low attogram level detection of alkaline phosphatase (AP)
  • Recommended for a wide range of AP applications including blotting (Western Blots, Southern blots), and for detection of alkaline phosphatase-labeled nucleotide and protein probes in solution and on solid supports such as ELISAs
  • 100X more sensitive than competitor products
  • Highest signal to noise ratios
  • High stability plus a 24 month shelf life

Specifications

Quantity 100 mL
For Use With (Application) Western Blot, ELISA, Southern Blot
Product Line Novus
Packaging Quantity 100 mL
Content And Storage Store at 4°C in the dark
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.