Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NovaLume Femto Chemiluminescent Substrate (HRP), ELISA
SDP

Catalog No. NBP261917 Shop All R&D Systems Products
Change view
Click to view available options
Packaging Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
NBP2-61917 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61917 Supplier Novus Biologicals™ Supplier No. NBP261917

NovaLume Femto Chemiluminescent Substrate (HRP) may be used for any ELISA application utilizing horseradish peroxidase (HRP) conjugates. The substrate can be used with various blocking buffers. Detection and analysis requires a chemiluminometer.

Chemiluminescent substrate supplied as two components:

  • High signal intensity and low background
  • High sensitivity - low femtogram level detection

Specifications

Quantity 200 mL
For Use With (Application) ELISA
Packaging Quantity 200 mL
Content And Storage Store at 4°C in the dark
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.