Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NovaLume Pico Chemiluminescent Substrate (HRP)
SDP

Catalog No. NBP261915 Shop All Novus Products
Change view
Click to view available options
Packaging Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
NBP2-61915 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61915 Supplier R&D Systems™ Supplier No. NBP261915

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. NovaLume Pico Chemiluminescent Substrate may be used for any blotting application utilizing horseradish peroxidase (HRP)-conjugates. The substrate can be used with various blocking buffers and on nitrocellulose or PVDF membranes. Visualization is by CCD imaging systems or x-ray film.

Chemiluminescent substrate supplied as two components:

  • High sensitivity - low pico to mid femtogram level detection
  • Recommended for HRP blotting applications
  • Superior performance to Competitor Products in its class

Specifications

Quantity 200 mL
For Use With (Application) Western Blot, Immunoblot, Dot Blot
Product Line Novus
Packaging Quantity 200 mL
Content And Storage Store at 4°C in the dark
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.