Learn More
PIP™ NOVAX™ Class 0 Electrical Safety Gloves Kit
Shop All Protective Industrial Products Inc. ProductsDescription
- Includes one pair of natural rubber electrical insulating gloves, a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag
- Class 0 insulating gloves will protect workers from up to 1000 V
- Meets NFPA 70E standards for use around arc flash hazards
Specifications
Specifications
| Certifications/Compliance | ANSI/ASTM D120, Class 0, Type I, NFPA 70E |
| Applications | Industrial |
| Color | Black |
| Hand | Specific |
| Material | Natural Rubber, Top Grain Goatskin Leather |
| Product Type | Electrical Insulating Gloves Kit |
| Size | 10 |
| For Use With (Application) | Electrical Utility Work, Industrial Plant Maintenance, and Power Generation |
| Glove Cuff Style | Straight |
| Includes | Natural Rubber Electrical Insulating Gloves (150-0-11), Top Grain Goatskin Leather Glove Protectors (148-1000), and a Nylon Glove Bag (148-2136) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.