Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Denaturing Cell Extraction Buffer

Catalog No. FNN0091
Encompass_Preferred
Click to view available options
Quantity:
100 ml

Suitable for use in ELISA, Western blotting, sample/assay prep

Denaturing Cell Extraction Buffer can be used for Invitrogen phosphoELISAs (i.e., ERK, JNK) that require sample treatment or boiling. After lysing cells with Denaturing Cell Extraction Buffer, lysates will not require sample treatment or boiling.

Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Product Type Cell Extraction Buffer
Quantity 100 ml
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.