Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EKMax™ Enterokinase

Catalog No. p-4925974
Encompass_Preferred
Click to view available options
Quantity:
1000 Units
250 Units

Includes

EKMax Enterokinase is provided with 10X buffer.

Recombinant preparation of the catalytic subunit of bovine enterokinase

  • Recognizes the sequence Asp-Asp-Asp-Asp-Lys and cleaves peptide bond after lysine residue
  • Enzyme can be used to cleave any fusion protein that carries this peptide sequence
  • Well-suited to removal of fusion tags from recombinant proteins
  • Unit Definition: One unit of EKMaxis amount of enzyme required to digest 20μg of thioredoxin-CAT fusion protein to 90% completion in 16 hours at 37°C; One EKMaxunit is equivalent to ∽190 trypsinogen activation units

GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Product Type Enterokinase
Content And Storage 250 or 1000 units of EKMax™ is provided with 10X buffer. Store at -20°C. Guaranteed stable for 6 months when stored properly.
Species Cattle/Bovine
Quantity 250 Units
Product Line EKMax™
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.