Learn More
Invitrogen™ InVision™ His-Tag In-Gel Staining Kit

Description
Includes
InVision™ His-tag In-gel Stain is supplied as a 1X ready-to-use staining reagent (500mL). BenchMark™ His-tagged Protein Standard (125 μL) is also included in the kit.
The InVision™ His-tag In-gel Staining Kit includes InVision™ His-tag In-gel Stain (also available separately), as well as BenchMark™ His-tagged Protein Standard. InVision™ His-tag In-gel Stain is a ready-to-use, fast, sensitive and highly specific stain for visualizing His-tagged fusion protein bands directly in a polyacrylamide gel following electrophoresis.
- Stain proteins fused to oligohistidine sequence
- Eliminate western blotting-detect His-tagged proteins directly in the gel
- Detect nanogram levels of protein bands with UV transilluminator, equipped with standard camera, or laser-based scanner
- Visualize results in under 3 hours
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Specifications
Specifications
Content And Storage | InVision™ His-tag In-gel Stain is supplied as a 1X ready-to-use staining reagent (500 mL). BenchMark™ His-tagged Protein Standard (125 μL) is also included in the kit. |
Detection Location | In-Gel Detection |
Detection Method | Fluorescence |
Label or Dye | InVision His-Tag Stain |
Product Type | His Tag Proteins In-Gel Staining Kit |
Product Line | InVision™ |
Quantity | 1 kit |
Target Molecule | Proteins (His-Tagged) |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.