Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, 8.6 x 13.5 cm

Catalog No. LC2006
Encompass_Preferred
Change view
Click to view available options
Quantity:
16 membrane/filter paper sandwiches
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2006 16 membrane/filter paper sandwiches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2006 Supplier Invitrogen™ Supplier No. LC2006
Add to Cart
Edge
Add to Cart

Fits 8.6 x 13.5 cm E-PAGE™ Gels.

 

  • Designed for western transfer of proteins
  • Pre-cut, extra-thick, absorbent Blotting Paper (2.5mm thickness) for E-PAGE
  • Pore Size: 0.45μm
  • Size: 8.6 x 13.5cm

 

A protein's properties affect its ability to bind to membrane surfaces.

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 13.5 cm
Width (Metric) 8.6 cm
Description Nitrocellulose/Filter Paper Sandwich, 0.45μm pore size
Quantity 16 membrane/filter paper sandwiches
Format Sandwich
Material Nitrocellulose
Pore Size 0.45 μm
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Dimensions (LxW) 8.6 cm x 13.5 cm
Thickness 2.5 mm
Product Line Novex
Show More Show Less
How should nitrocellulose be treated prior to transferring proteins?

Prewet the nitrocellulose membrane in distilled water or in 1X transfer buffer for 5 min with gentle shaking. This is to prevent any dry spots in the membrane that may interfere with transfer.

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.