Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Tau (Total) ELISA Kit

Catalog No. p-4926686
Encompass_Preferred
Click to view available options
Quantity:
2 x 96 Tests
96 Tests

Includes

Pre-coated plate, standard, biotinylated detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

ELISA

The Human Tau (Total) ELISA quantitates Hu Tau in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Tau, independent of phosphorylation state. Principle of the method The Human Tau solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10636
Assay Range 31 to 2000 pg/mL
Assay Sensitivity <10 pg/mL
Concentration 31 to 2000 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cerebrospinal Fluid, Supernatant
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4137
Gene Symbol DDPAC,FTDP-17 L,MSTD,MTBT1,MTBT2,PPND,PPP1R103,TAU,MAPT
Incubation Time 4 hr.
Interassay CV 6.5%
Intraassay CV 4.6
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Tau
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Cerebrospinal Fluid 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Tau
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.