Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Prostaglandin E2 ELISA Kit

Catalog No. KHL1701
Encompass_Preferred
Click to view available options
Quantity:
96 Tests

ELISA

The Human Prostaglandin E2 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (ELISA). This assay is designed to detect and quantify the level of human prostaglandin E2 in biological samples. Principle of the method This assay is based on the competition between Prostaglandin E 2 (PGE2) and a PGE2 alkaline phosphatase tracer for a limited amount of PGE2 specific monoclonal antibody. Because of the competition between PGE2 in the sample and PGE2 tracer for the PGE2 antibody, the signal obtained with the assay will be inversely proportional to the amount of PGE 2 in each sample. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Prostaglandin E2 (PGE2) is a lipid mediator derived from arachidonic acid through the actions of cyclooxygenase (COX) enzymes and terminal prostaglandin E synthases (PGES). It plays a significant role in mediating inflammation, pain sensation, and fever by acting on four receptor subtypes: EP1, EP2, EP3, and EP4. PGE2's involvement in inflammatory responses is well-documented, particularly in diseases such as rheumatoid arthritis and osteoarthritis. Various medications, including nonsteroidal anti-inflammatory drugs (NSAIDs) and selective COX-2 inhibitors, are used to reduce PGE2 production, thereby alleviating inflammation and pain. However, these drugs can have side effects, such as gastrointestinal issues and increased risk of thrombosis. Additionally, PGE2 has a critical role in immune regulation, affecting T cell function and, by extension, the adaptive immune response. It also has a significant impact on skeletal disorders and pain through mechanisms that are still being explored.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM
Assay Range 31.3 to 4000 pg/mL
Conjugate AP
Product Type ELISA
Sample Type Plasma, Supernatant, Urine
Target Species Human
For Use With (Application) Colorimetric Detection
For Use With (Equipment) Colorimetric Microplate Reader
Incubation Time 3 hr.
Interassay CV 11.7%
Intraassay CV 6
Kit Contents Pre-coated 96 well plate, Standard, Tris Buffer, Alkaline Phosphatase Tracer, Leukotriene B4 Antiserum, DEA Buffer, Wash Buffer, pNPP tablets, Adhesive Plate Covers
Label or Dye AP
Marker Gene Prostaglandin E2
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Supernatant 100 μL, Urine 100 μL
Storage Requirements Store kit at -20°C. Refer to product documentation for component specific storage temperature.
Target PGE2
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.