Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ TBE Gels, 20%

Catalog No. p-7250383
Encompass_Preferred
Click to view available options
Wells:
10-well
12-well
15-well

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels.

Novex™ TBE Gels 20% designed to run on the XCell SureLock™ Mini-Cell provide high-resolution analysis of restriction digests and PCR products.

Novex™ TBE Gels 20% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 15–50 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium­ bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:
• High resolution and sensitivity with lower background staining
• Requires ∼10% sample concentration and volume of large or agarose gels
• Efficient blotting
• Easy extraction of DNA from gels
• Gel extraction does not interfere with enzymatic reactions
• Accurate and reproducible results

Formulation
Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Catalog number refers to a single box of 10 gels

Store at 4°C.
Description Novex TBE Gels, 20%, 10-well
For Use With (Equipment) XCell SureLock Mini-Cell
Gel Percentage 20%
Gel Type TBE
Product Type TBE Gel
Separation Range 15 to 50 bp
Wells 10-well
Quantity 10 Gels/Box
Sample Type Double-stranded DNA (dsDNA)
Shipping Condition Wet Ice
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.