Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novobiocin Sodium Salt, MP Biomedicals™

Catalog No. MP215595780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215595780 100 g
MP215595701 1 g
MP215595705 5 g
MP215595725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215595780 Supplier MP Biomedicals, Inc Supplier No. 0215595780

Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II. Novobiocin Sodium Salt, MP Biomedicals is an antibiotic produced by Streptomyces nieveus. It is active against gram-positive bacteria. It is also a Hsp90 inhibitor 3.

Novobiocin inhibits DNA synthesis by inhibiting bacterial DNA gyrase targeting the GyrB subunit of the enzyme involved in energy transduction. It acts as a competitive inhibitor of the ATPase reaction catalysed by GyrB 2. Uses include:

  • For the production of positively supercoiled plasmid DNA
  • Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase
  • Inhibitor of retrovirus RNA-dependent DNA-polymerase
  • To study heat shock protein inhibition
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-53-5
Molecular Formula C31H35N2NaO11
Molecular Weight (g/mol) 634.61
MDL Number MFCD00066541,MFCD00066541
InChI Key AXOUUAINTJNFRS-UHFFFAOYNA-N
Synonym Albamycin, Cathomycin sodium
PubChem CID 131673945
SMILES [Na+].COC1C(OC(N)=O)C(O)C(OC2=CC=C3C(=O)[C-](NC(=O)C4=CC=C(O)C(CC=C(C)C)=C4)C(=O)OC3=C2C)OC1(C)C

Specifications

Physical Form Powder
Absorbance λ max: 307nm
Melting Point 220°C
Extinction Coefficient E1%= 600 (0.1M NAOH)
Potency ∼900μg/mg
Quantity 100 g
Optical Rotation −38.0° (2.5g/100ml ethanol at 24°C for the free acid)
Solubility Information Freely soluble in water (10%, clear, colorless solution)
IUPAC Name sodium 7-{[4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy}-3-[4-hydroxy-3-(3-methylbut-2-en-1-yl)benzamido]-8-methyl-2,4-dioxo-3,4-dihydro-2H-1-benzopyran-3-ide
Formula Weight 634.6
Recommended Storage Store at 2–8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.