Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 3363 |
| Species | Human |
| Accession Number | P34969 |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-46598. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 |
| Gene Symbol | HTR7 |
| Product Type | 5-HT7 |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
