Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ AASD-PPT Protein
SDP

Catalog No. NBP14930701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14930701 0.1 mg
NBP1493075 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14930701 Supplier Novus Biologicals™ Supplier No. NBP1493070.1MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 14-309 of Human AASD-PPT The Recombinant Human AASD-PPT Protein is derived from E. coli. The Recombinant Human AASD-PPT Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl Buffer (pH 8.0) containing 1mM DTT, 10% Glycerol
Gene ID (Entrez) 60496
Molecular Weight (g/mol) 36.4kDa
Purification Method Protein
Quantity 0.1 mg
Source Human
Immunogen AASDHPPT, 14-309aa. Sequence: MGSSHHHHHHSSGLVPRGSHMEGVRWAFSCGTWLPSRAEWLLAVRSIQPEEKERIGQFVFARDAKAAMAGRLMIRKLVAEKLNIPWNHIRLQRTAKGKPVLAKDSSNPYPNFNFNISHQGDYAVLAAEPELQVGIDIMKTSFPGRGSIPEFFHIMKRKFTNKEWETIRSFKDEWTQLDMFYRNWALKESFIKAIGVGLGFELQRLEFDLSPLNLDIGQVYKETRLFLDGEEEKEWAFEESKIDEHHFVAVALRKPDGSRHQDVPSQDDSKPTQRQFTILNFNDLMSSAVPMTPEDPSFWDCFCFTEEIPIRNGTKS
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name AASD-PPT
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.