Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ABCA1 Antibody Blocking Peptide
SDP

Catalog No. NB400105PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400105PEP 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400105PEP Supplier Novus Biologicals™ Supplier No. NB400105PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A peptide to ABCA1. The ABCA1 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 19
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB 400-105. Use in Blocking/Neutralizing Was Reported in Scientific Literature (PMID: 24612239) For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Gene Symbol ABCA1
Product Type ABCA1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.