Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ACBP Protein
SDP

Catalog No. NBP1508335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1508335 0.5 mg
NBP15083301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1508335 Supplier Novus Biologicals™ Supplier No. NBP1508330.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-87 of Human ACBP The Recombinant Human ACBP Protein is derived from E. coli. The Recombinant Human ACBP Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001073331
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol
Gene ID (Entrez) 1622
Molecular Weight (g/mol) 12.2kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Human
Immunogen ACBP, 1-87aa, Human, His tag, E. coli (NP_001073331).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name ACBP
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.