Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 176 |
| Species | Mouse |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-74350. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN |
| Gene Symbol | ACAN |
| Product Type | Aggrecan |
| Quantity | 0.1 mg |
| Regulatory Status | RUO - research use only |
| Termination | Synthetic peptide corresponding to residues C G G N(aa 387) I T E G E(aa 392) of human aggrecan |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
