Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ AIF-1/Iba1 Protein

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-147 of Human AIF-1/Iba1 The Recombinant Human AIF-1/Iba1 Protein is derived from E. coli. The Recombinant Human AIF-1/Iba1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NP_001614 |
Concentration | 0.5mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 20% glycerol, 200mM NaCl |
Gene ID (Entrez) | 199 |
Molecular Weight (g/mol) | 18.9kDa |
Purification Method | SDS-PAGE |
Quantity | 0.5 mg |
Source | E. coli |
Immunogen | AIF1, 1-147aa, Human, His tag, E. coli (NP_001614). |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction