Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Antennapedia Control Recombinant Protein
SDP

Catalog No. NB81074573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB81074573 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB81074573 Supplier Novus Biologicals™ Supplier No. NB81074573

Highly purified. Generating reliable and reproducible results.

Recombinant Luciferase is a luciferase expressed from a cloned gene from the North American firefly (Photinus pyralis). The Recombinant Firefly Luciferase (firefly) Protein is derived from E. coli.

Specifications

Formulation 25mM Tris-acetate, pH 7.8 (4C), 1mM EDTA, 1mM DTT, 0.2M ammonium sulfate, 15% glycerol, and 30% ethylene glycol.
Name Luciferase Protein
Purification Method Protein
Quantity 0.5 mg
Source E.Coli
Immunogen Recombinant (Photinus pyralis)
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Firefly
Recombinant Recombinant
Research Category Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.