Learn More
Description
Specifications
Specifications
| Antigen | IgG Protein |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot, ELISA |
| Dilution | Flow Cytometry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, SDS-Page, Fluorophore-linked immunosorbent assay 1:100 - 1:2000 |
| Formulation | Potassium Phosphate, 0.15M Sodium Chloride pH 7.2. |
| Quantity | 1 mg |
| Content And Storage | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
