Learn More
Specifications
Specifications
| Gene ID (Entrez) | 1588 |
| Accession Number | P11511 |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Formulation | PBS [pH 7.2] containing 50% glycerol and 0.5% BSA |
| For Use With (Application) | For blocking the antibody activity of Aromatase (cat. #NBP1-45360). |
| Product Type | Aromatase |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
