Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Aromatase Antibody Blocking Peptide

Click to view available options
Quantity:
0.05 mg
Description
A peptide to CYP19A1.
Specifications
Specifications
Gene ID (Entrez) | 1588 |
Species | Human |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for BC100-1596. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450 AROM, Estrogen synthase, microsomal monooxygenase |
Gene Symbol | CYP19A1 |
Product Type | Aromatase |
Quantity | 0.05 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction