Learn More
Novus Biologicals™ Recombinant Human Aspartate beta hydroxylase His Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | NP_001158227 |
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol |
| Gene ID (Entrez) | 444 |
| Molecular Weight (g/mol) | 24.5kDa |
| Name | Aspartate beta hydroxylase Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MFDLVDYEEV LGKLGIYDAD GDGDFDVDDA KVLLGLKERS TSEPAVPPEE AEPHTEPEEQ VPVEAEPQNI EDEAKEQIQS LLHEMVHAEH ETEHSYHVEE TVSQDCNQDM EEMMSEQENP DSSEPVVEDE RLHHDTDDVT YQVYEEQAVY EPLENEGIEI TEVTAPPEDN PVEDSQVIVE EVSIFPVEEQ QEVPPDT |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.