Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Aurora B His Protein
SDP

Catalog No. NBP130242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130242 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130242 Supplier Novus Biologicals™ Supplier No. NBP130242
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1- 344 of Human Aurora B The Recombinant Human Aurora B Protein is derived from E. coli. The Recombinant Human Aurora B Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q96GD4
For Use With (Application) ELISA, Western Blot
Formulation 20mM Tris-HCl buffer (pH8.0), 0.5mM DTT, 20% glycerol, 0.1mM EDTA, 0.1mM EGTA, 0.1 M NaCl, 0.1mM PMSF
Gene ID (Entrez) 9212
Molecular Weight (g/mol) 41.4kDa
Name Aurora B Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Aurora Kinase B, 1- 344 aa, Human, His-tagged, Recombinant, E.coli (NP_004208).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.