Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Brk Protein
SDP

Catalog No. NBP199459
Click to view available options
Quantity:
0.005 mg

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant protein for Human Brk The Recombinant Human Brk Protein is derived from Sf 9 (baculovirus). The Recombinant Human Brk Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Accession Number NM_005975.
Formulation Recombinant protein in storage buffer (50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol).
Gene ID (Entrez) 5753
Molecular Weight (g/mol) 80kDa
Name Brk Protein
Purification Method SDS-PAGE
Quantity 0.005 mg
Source Baculovirus (Sf9 insect cells)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.