Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 6347 |
| Species | Rat |
| Purification Method | Peptide affinity purified |
| Purity | Immunogen affinity purified |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-07035. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF |
| Gene Symbol | CCL2 |
| Product Type | CCL2/MCP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
